Dropping bottles, LDPE, Nalgene®
Supplier: Thermo Fisher Scientific
2416-0060 2416-0125
NALG2416-0060EA 0 EUR
NALG2416-0060 NALG2416-0125
Dropping bottles, LDPE, Nalgene®
Bottles Dropping Bottles
LDPE bottle, PE dropping pipette, neoprene bulb.
  • Suitable for dispensing most acids, bases and alcohols
  • Good temperature resistance
  • Easy handling
Dispense at a rate you control, be it one drop at a time or in a stream, with LDPE bottles with dropper assembly.