PIG® vertical mat roll holder
Supplier: NEW PIG
GEN249
NEWPGEN249EA 260 EUR
NEWPGEN249
PIG® vertical mat roll holder
Spill Control Materials
No more lugging around large rolls of mat, let this vertical mat roll holder do all the work.
  • No-tip swivel wheel casters allow holder to move freely in any direction
  • Constructed of powder-coated, hot-rolled steel with a comfortable handle
  • 8 cm high platform is low enough for easy mat positioning
For use with any PIG mat roll up to 93 cm width x 51 cm diameter, holds up to 41 kg.