PIG® Quick-Response High-Visibility Spill Carts
Supplier: NEW PIG
KIT2401-999 KIT3401-999
NEWPKIT2401-999EA 636.5 EUR
NEWPKIT2401-999 NEWPKIT3401-999
PIG® Quick-Response High-Visibility Spill Carts
Spill Control Materials
Get to the spill fast and fully stocked with easy-to-access absorbents in this mobile, high-visibility yellow spill kit.
  • Absorb anything from water or coolants to oil or solvents with the pre-packed absorbents inside this spill caddy
  • Manoeuvre effortlessly, even through doorways, thanks to the tip-back design and snag-resistant 18 cm wheels
  • Contents are compartmentalised for easy access
Easily work to clean any small- or moderate-sized spill since the spill caddy's spacious interior holds 75 L worth of absorbents and spill-response items, and theMore Product Information