Elephant mat, zigzag stitched PIG®
Supplier: NEW PIG
MAT234 MAT256 MAT270 MAT286
NEWPMAT234EA 263.75 EUR
NEWPMAT234 NEWPMAT256 NEWPMAT270 NEWPMAT286
Elephant mat, zigzag stitched PIG®
Spill Control Materials
The zigzag stitched PIG® mat is made from eight-layered, highly absorbent, fine-fibre 100% PP construction. The zigzag stitch offers excellent walk-on durability. It absorbs most common industrial liquids like oils, water, solvents, coolants.
  • Dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Flame-resistant - material melts when exposed to high heat
  • Can be incinerated after use
The mat is designed specifically for heavy foot traffic or moderate to light wheeled traffic. Rolls are ideal for walkways and large areas toMore Product Information