PIG® acid solidifying absorbent powder
Supplier: NEW PIG
PLP501
NEWPPLP501EA 263 EUR
NEWPPLP501
PIG® acid solidifying absorbent powder
Spill Control Materials
Solidifies absorbed acids and locks them into the material for easier cleanup.
  • Permanently solidifies to "lock in" absorbed liquids; won't leak or revert to liquid state when handled or placed under pressure
  • Forms a solid mass without expanding for easier, less expensive recovery and cleanup; may help you comply with EPA requirements when used to solidify waste prior to shipment or landfilling
  • High Btu value produces less than 0.1% ash content when burned; ideal for fuels blending and incineration
  • Shaker bottle makes it easy to apply absorbent directly