Crucible tongs with bow, stainless steel
Supplier: NICKEL ELECTRO
3060 3061 3063 3064 3066
NICK3060EA 10.7 EUR
NICK3060 231-0357 NICK3063 NICK3064 231-0358
Crucible tongs with bow, stainless steel
Tongs
Stainless steel 18/10, anti-magnetic. For easy, safe handling of crucibles made from a wide variety of materials.
  • With grooved tips
  • Excellent corrosion resistance
  • Good thermal and electrical conductivity
  • Excellent thermal resistance