Bowls, evaporating, with spout
Supplier: NICKEL ELECTRO
715
NICK715EA 38.3 EUR
NICK715
Bowls, evaporating, with spout
Trays/Dishes/Pots Evaporating Basins/Dishes
Pure nickel (99,6%).
  • Excellent corrosion and temperature resistance
  • Smooth surfaces for fast, easy cleaning
Round, with flat bottom. Multipurpose tray ideal for a wide variety of uses.