Anti-KRT1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: LHK1]

Supplier: Novus Biologicals
NB100-2756H
NOVUNB100-2756HEA 758 EUR
NOVUNB100-2756H
Anti-KRT1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: LHK1]
Antibodies
Type: Primary
Antigen: KRT1 (keratin 1)
Clonality: monoclonal
Clone: LHK1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR