Anti-KAT3A/CBP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-381
NOVUNB100-381EA 706 EUR
NOVUNB100-381
Anti-KAT3A/CBP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal KAT3A/CBP Antibody. Tested Applications: Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation, Proximity Ligation Assay. Tested Reactivity: Human.

There are 2 scientific publications using this antibody.

Type: Primary
Antigen: KAT3A/CBP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR