Anti-TUBA Mouse Monoclonal Antibody (DyLight® 488) [clone: DM1A]

Supplier: Novus Biologicals
NB100-690G
NOVUNB100-690GEA 650 EUR
NOVUNB100-690G
Anti-TUBA Mouse Monoclonal Antibody (DyLight® 488) [clone: DM1A]
Antibodies
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: DM1A
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR