Anti-BECN2 Rabbit Polyclonal Antibody (DyLight® 650)

Supplier: Novus Biologicals
NB110-60982C
NOVUNB110-60982CEA 668 EUR
NOVUNB110-60982C
Anti-BECN2 Rabbit Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Primary
Antigen: BECN2 (beclin 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol BECN2
Antigen name Beclin 2
Environmentally Preferable
Antigen synonyms BECN1P1|BECN1L1
Antibody type Primary
Clonality Polyclonal
Conjugation DyLight® 650
Reactivity Human
Host Rabbit
Gene ID 441925
Isotype
Western blot Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR