Anti-LMO2 Mouse Monoclonal Antibody (DyLight® 488) [clone: 1A9-3B11]

Supplier: Novus Biologicals
NB110-78626G
NOVUNB110-78626GEA 722 EUR
NOVUNB110-78626G
Anti-LMO2 Mouse Monoclonal Antibody (DyLight® 488) [clone: 1A9-3B11]
Antibodies
Type: Primary
Antigen: LMO2 (LIM domain only 2 (rhombotin-like 1))
Clonality: monoclonal
Clone: 1A9-3B11
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR