Anti-MYC Mouse Monoclonal Antibody [clone: 9E11]

Supplier: Novus Biologicals
NB200-108 NB200-108SS
NOVUNB200-108EA 667 EUR
NOVUNB200-108 NOVUNB200-108SS
Anti-MYC Mouse Monoclonal Antibody [clone: 9E11]
Antibodies
The c-Myc Antibody (9E11) from Novus Biologicals is a mouse monoclonal antibody to c-Myc. This antibody reacts with human, mouse, chicken, yeast. The c-Myc Antibody (9E11) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: 9E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR