Anti-Neurokinin 1 Receptor Rabbit Polyclonal Antibody (DyLight® 650)

Supplier: Novus Biologicals
NB300-119C
NOVUNB300-119CEA 800 EUR
NOVUNB300-119C
Anti-Neurokinin 1 Receptor Rabbit Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Primary
Antigen: Neurokinin 1 Receptor
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR