Anti-CD59 mouse monoclonal antibody [clone: MEM-43]

Supplier: Novus Biologicals
NB500-330
NOVUNB500-330EA 542 EUR
NOVUNB500-330
Anti-CD59 mouse monoclonal antibody [clone: MEM-43]
Antibodies
The CD59 Antibody (MEM-43) from Novus Biologicals is a mouse monoclonal antibody to CD59. This antibody reacts with human. The CD59 Antibody (MEM-43) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Blocking / Neutralising, CyTOF-ready.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR