Anti-HSPA5 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NBP1-06277B
NOVUNBP1-06277BEA 722 EUR
NOVUNBP1-06277B
Anti-HSPA5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR