Anti-Oval Cell Marker Rat Monoclonal Antibody (Biotin) [clone: OC2-6E10]

Supplier: Novus Biologicals
NBP1-18981B
NOVUNBP1-18981BEA 668 EUR
NOVUNBP1-18981B
Anti-Oval Cell Marker Rat Monoclonal Antibody (Biotin) [clone: OC2-6E10]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-6E10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR