Anti-EIF4B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-24475
NOVUNBP2-24475EA 434 EUR
NOVUNBP2-24475
Anti-EIF4B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EIF4B (eukaryotic translation initiation factor 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR