Anti-S100A8 Rat Monoclonal Antibody [clone: 63N13G5]

Supplier: Novus Biologicals
NBP2-27067
NOVUNBP2-27067EA 434 EUR
NOVUNBP2-27067
Anti-S100A8 Rat Monoclonal Antibody [clone: 63N13G5]
Antibodies
Type: Primary
Antigen: S100A8 (S100 calcium binding protein A8)
Clonality: monoclonal
Clone: 63N13G5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR