Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE-1/AE-3]

Supplier: Novus Biologicals
NBP2-29429
NOVUNBP2-29429EA 0 EUR
NOVUNBP2-29429
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE-1/AE-3]
Antibodies
Type:
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: AE-1/AE-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Rabbit, Reptile, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR