Orange G, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

O7252-100G O7252-25G
O7252-100GEA 273 EUR
O7252-100G O7252-25G
Orange G, Sigma-Aldrich®
Orange G
Formula: C₁₆H₁₀N₂Na₂O₇S₂
Melting Pt: 141 °C
Density: 0.45…0.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db9cc20a6cb713d4babe5cf40f4fb3f2350cec65&vid=69ba3e383bb0d5764034aeb0289056e797456d3a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR