Guardian 7000 magnetic hotplate stirrers
Supplier: OHAUS
30500603 30500613 30500623
OHAU30500603EA 1050 EUR
OHAU30500603 OHAU30500613 OHAU30500623
Guardian 7000 magnetic hotplate stirrers
Stirrers Magnetic Hotplate Stirrers
Durable magnetic hotplate stirrers with superior safety features and intelligent performance.
  • SmartPresence™ and SmartLink™ technologies allow unsupervised operation
  • SafetyHeat™ system protects the laboratory
  • Powerful, consistent stirring performance
  • Customised performance for precise applications
OHAUS Guardian 7000 magnetic hotplate stirrers are designed with outstanding safety features and superior heating and mixing performance. The SmartHousing™ is chemical resistant with an innovative, easy-to-clean design that channels spills away from internalMore Product Information