Endeavor™ Microplate Shaker
Supplier: OHAUS
30770917
OHAU30770917EA 0 EUR
OHAU30770917
Endeavor™ Microplate Shaker
Shakers Microplate Shakers
Stable, long-term shaking with triple eccentric drive and maintenance-free brushless DC motor.
  • Adaptable to a wide variety of accessories to customize for low to mid volume applications
  • Programmable with a simple on-board menu to save time and guarantee reproducible results
  • Remote function permits PC control and data transfer
For use in a wide variety of applications including microbiology, molecular biology, cell cultures, staining/destaining, DNA studies, environmental and food sciences, chemistry, general mixing and more.
Feature a bright,