Accessories for Moxi™ Z

Supplier: Orflo
MXA004 MXC011 MXC012
701-0252EA 0 EUR
701-0252 ORFLMXC011 ORFLMXC012
Accessories for Moxi™ Z
Cell Counters and Flow Cytometers Automated Cell Counters
Order Now

SPECIFICATIONS

Model Type M cassettes Type S cassettes
Environmentally Preferable
Concentration 3000 – 500000 cells/ml 3000 – 1750000 cells/ml
Cell size 34 - 20580 fL 14 - 9202 fL
Sample volume 75 µl 75 µl
Dynamic range 4 - 34 µm 3 - 26 µm
Measurement time 8 s 15 s

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR