DH5α Chemically Competent E. coli Cells
Supplier: OriGene
CC100003
ORIGCC100003EA 223 EUR
ORIGCC100003
DH5α Chemically Competent E. coli Cells
Competent Cells
  • lacZ marker for blue/white colour screening
  • Transformation of cloned DNA into bacterial cells
  • Ideal for subcloning and generating cDNA libraries
Kit contains:
DH5α chemically competent cells (5×200 μl)
Positive control DNA - pUC19 (50 pg/μl, 25 μl)
SOC medium: 20 ml