Anti-ERG Mouse Monoclonal Antibody [clone: UMAB77]

Supplier: OriGene
UM570065
ORIGUM570065EA 311 EUR
ORIGUM570065
Anti-ERG Mouse Monoclonal Antibody [clone: UMAB77]
Antibodies
Recommended Dilutions: Immunochemistry: 1:100

Type: Primary
Antigen: ERG
Clonality: Monoclonal
Clone: UMAB77
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR