Accessories for Orion™ pH Electrodes
Supplier: Thermo Orion
910005 910006-WA
ORIO910005EA 43.8 EUR
ORIO910005 ORIO910006-WA
Accessories for Orion™ pH Electrodes
Electrodes pH Electrodes
Accessories for storing and protecting pH electrodes. Storage bottles protect and condition electrodes when not in use; bulb guard protects the pH sensing bulb of glass pH electrodes.
  • Electrode storage bottles can be used with any 6, 8 or 12 mm pH electrodes
  • Electrode bulb guard: Designed for glass body pH electrodes; for use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes
  • Bulb guard: One end of the bulb guard fits over standard electrodes while the other end fits over sure-flow electrodes without interfering with the sure-flow flushing action