Sequencing auxiliary vials
Supplier: Oxford Nanopore Technologies
EXP-AUX003
OXNTEXP-AUX003EA 105 EUR
OXNTEXP-AUX003
Sequencing auxiliary vials
Nucleic Acid Reagents Next Generation Sequencing Reagents
Additional sequencing buffer, elution buffer, and loading beads.
These kits are recommended for users who would like to split a library across multiple flow cells, need to add more library/a new library onto a washed flow cell, or wish to top up a flow cell with additional library.
Sequencing Auxiliary Vials (EXP-AUX003) contain Sequencing Buffer (SB), Elution Buffer (EB), Library Solution (LIS) and Library Beads (LIB) with v14 flow cell priming reagents: Flow Cell Flush (FCF) and Flow Cell Tether (FCT). This product is compatible with R10.4.1 Flow Cells and all kits using Kit 14 chemistry (e.g. Ligation Sequencing Kit SQK-LSK114).
Delivery information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance. Oxford Nanopore products are only available within European Union countries