Rapid Adapter Auxiliary V14
Supplier: Oxford Nanopore Technologies
EXP-RAA114
OXNTEXP-RAA114EA 305 EUR
OXNTEXP-RAA114
Rapid Adapter Auxiliary V14
Nucleic Acid Reagents Next Generation Sequencing Reagents
Additional rapid adapter and adapter buffer to enable customers to maximise the use of rapid barcoding kits 24 and 96 V14 (SQK-RBK114.24 and SQK-RBK114.96).
The rapid adapter auxiliary V14 (EXP-RAA114) expansion contains additional rapid adapter and adapter buffer for another six reactions. This is to enable customers to use any unused barcodes in their rapid barcoding kits V14.
To use this kit, ensure to purchase the sequencing auxiliary vials V14 (EXP-AUX003) to provide the flow cell priming reagents.
Delivery information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance. Oxford Nanopore products are only available within European Union countries