MinION™ Mk1D DNA/RNA Sequencing Device
Supplier: Oxford Nanopore Technologies
SUPP1D MDP-RPB114 BASIC12M-MK1D MDIP-LSK114 MDIP-NBD14.24 MDIP-PCS14 MDIP-RAD14 MDIP-RBK14.24 MDIP-RNA MDIP-RPB114 MDP-LSK114 MDP-NBD14.24 MDP-PCS14 MDP-RAD14 MDP-RBK14.24 MDP-RNA
OXNTSUPP1DEA 6270 EUR
OXNTSUPP1D OXNTMDP-RPB114 OXNTBASIC12M-MK1D OXNTMDIP-LSK114 OXNTMDIP-NBD14.24 OXNTMDIP-PCS14 OXNTMDIP-RAD14 OXNTMDIP-RBK14.24 OXNTMDIP-RNA OXNTMDIP-RPB114 OXNTMDP-LSK114 OXNTMDP-NBD14.24 OXNTMDP-PCS14 OXNTMDP-RAD14 OXNTMDP-RBK14.24 OXNTMDP-RNA
MinION™ Mk1D DNA/RNA Sequencing Device
Next Generation Sequencing Instruments
MinION™ - your personal, portable DNA and RNA sequencer.
  • Any length read from short to ultra long: 20 bp to >4 Mb
  • High-accuracy, direct sequencing of native DNA and RNA with built-in modification and best-in-class methylation detection
  • Real-time sequencing for immediate access to actionable results
  • Simple 10-minute sample prep available
  • Portable, pocket-sized device sequence in the field or lab
A sequencer for everyone - empowering individual researchers, labs and those new to DNA/RNA sequencing to perform in-house sequencing and take control ofMore Product Information
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance. Oxford Nanopore products are only available within European Union countries