GridION™ DNA/RNA Benchtop Nanopore Sequencer
Supplier: Oxford Nanopore Technologies
SUPP001 GRD-MK1CAPX SUPP006 BASIC12M-G BASIC24M-G BASIC36M-G
OXNTSUPP001EA 15660 EUR
OXNTSUPP001 OXNTGRD-MK1CAPX OXNTSUPP006 OXNTBASIC12M-G OXNTBASIC24M-G OXNTBASIC36M-G
GridION™ DNA/RNA Benchtop Nanopore Sequencer
Next Generation Sequencing Instruments
GridION™ - a self-contained, easily deployable DNA/RNA benchtop nanopore sequencer.
  • Flexible - run up to five independently addressable flow cells
  • Integrated compute - powerful onboard data processing and analysis, minimizing IT requirements
  • Compact - small footprint, suitable for any lab
  • Real-time data analysis - rapid results with base modification data as standard
  • Any read length - from short to ultra-long (longest >4 Mb)
A flexible, self-contained, benchtop nanopore sequencer, running up to five MinION™ Flow Cells that can respond to the needs
More Product Information
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance. Oxford Nanopore products are only available within European Union countries