Hype-5 transfection blast is dedicated to achieve high yield protein expression in mammalian cells. This product has been designed f or maximum recombinant protein expression in hek293 and cho cells growing in suspension. 1 * 5 mL
Supplier: OZ BIOSCIENCES
Hype-5 transfection blast is dedicated to achieve high yield protein expression in mammalian cells. This product has been designed f or maximum recombinant protein expression in hek293 and cho cells growing in suspension. 1 * 5 mL