PDAM, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: Pyrene, 1-(diazomethyl), 1-Pyrenyldiazomethane

P6863-25MG
P6863-25MGEA 661 EUR
P6863-25MG
PDAM, Sigma-Aldrich®
PDAM

Fluorescent diazoalkane useful as an HPLC derivatization reagent for carboxylic acid detection with better detection limit parameters and greater chemical stability than the commonly used 9-anthryldiazomethane.

Formula: C₁₇HN₂
MW: 242.27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467268
CAS Number: 78377-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc707a123c83333ebc87df7760af7a3923403b3&vid=c6caf7f1d7c0c6927e61a90d98fa12803119dda0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR