Human plasma, chronic myeloid leukemia (CML)
Supplier: Charles River Laboratories Cell Solutions
PB006C-1-CML
PB006C-1-CMLEA 0 EUR
PB006C-1-CML
Human plasma, chronic myeloid leukemia (CML)
Plasma
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of chronic myeloid leukemia (CML). Collected from IRB consented chronic myeloid leukemia (CML) disease donors.
No pre screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Caution: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.