Nebulisers, PFA-ST/ST3 MicroFlow™
Supplier: PerkinElmer
N8122192 N8145101 N8145368
PERCN8122192EA 1680 EUR
PERCN8122192 PERCN8145101 PERCN8145368
Nebulisers, PFA-ST/ST3 MicroFlow™
Atomisers
The PFA-ST MicroFlow nebuliser is made from high purity, HF resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the diameter of the external capillary or probe.
  • Chemically resistant ideal for strong acids, alkalis and organics
  • Direct analysis of volatile and non-volatile organic solvents
  • Longer lifetime than glass or quartz nebulisers
  • With external 1/4 - 28 threaded connector
Exchangeable uptake capillaries allow one nebuliser to be used at different self-aspiration rates from 20 to 700 μl/More Product Information