Molybdenum Standard Solution,1,000 μg/ml, 2% HNO₃
Supplier: PerkinElmer
N9303745
PERCN9303745EA 82.8 EUR
PERCN9303745
Molybdenum Standard Solution,1,000 μg/ml, 2% HNO₃
Standards Cationic Standards Molybdenum Standards
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Caution: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).