SPE columns, Supra-Clean® Florisil (FL-S)
Supplier: PerkinElmer
N9306413 N9306494 N9306513 N9306514 N9306515
PERCN9306413EA 137 EUR
PERCN9306413 PERCN9306494 PERCN9306513 PERCN9306514 PERCN9306515
SPE columns, Supra-Clean® Florisil (FL-S)
Chromatography Columns
Supra-Clean® Florisil (FL-S) SPE products are recommended for extracting polar compounds when analysing pesticides and polychlorinated biphenyls (PCB).
  • Standard grade
  • Hydrophilic mechanism
  • Ideal for polar compounds in non-polar matrix
  • pH range: 2 to 8
Alternative to silica for viscous matrices due to large particle size.