Inclusion body solubilisation reagent, Pierce™
Supplier: Thermo Fisher Scientific
78115
PIER78115EA 293 EUR
PIER78115
Inclusion body solubilisation reagent, Pierce™
Protein Purification Protein Purification Reagents
Inclusion Body Solubilisation Reagent is designed to retrieve expressed protein in soluble form after lysis and extraction with reagents.
  • Achieves the essential first step for subsequent insoluble protein refolding procedures
  • One easy to use solution
  • Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE may be performed
  • Can accommodate additional components such as reducing agents or chelating agents