n-Dodecyl-β-D-maltoside

Supplier: Thermo Fisher Scientific
Warning

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside

89902 89903
PIER89902EA 179 EUR
PIER89902 PIER89903
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
Storage Temperature: Freezer
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR