5-Chloro-2-(methylamino)benzophenone

Supplier: LGC Standards PROMOCHEM

ICRS0199
PROOICRS0199EA 0 EUR
PROOICRS0199
5-Chloro-2-(methylamino)benzophenone
5-Chloro-2-(methylamino)benzophenone
Formula: C₁₄H₁₂ClNO
MDL Number: MFCD00008284
CAS Number: 1022-13-5
EINECS: 213-822-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR