Naphthalene

Supplier: LGC Standards PROMOCHEM
Warning

LGC2603
PROOLGC2603EA 496 EUR
PROOLGC2603
Naphthalene
Naphthalene
Formula: C₁₀H₈
MW: 128.17 g/mol
Boiling Pt: 218
Melting Pt: 79…82 °C
Density: 1.145 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Ambient
MDL Number: MFCD00001742
CAS Number: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1 III
Merck Index: 12,06457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR