Isonicotinamide

Supplier: LGC Standards PROMOCHEM

Synonyms: 4-Pyridinecarboxylic acid amide

MM0552.01
PROOMM0552.01EA 1290 EUR
PROOMM0552.01
Isonicotinamide
Isonicotinamide
Formula: C₆H₆N₂O
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 155…157 °C
MDL Number: MFCD00006432
CAS Number: 1453-82-3
EINECS: 215-926-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR