Meat homogenate - nutrients, NIST-1546A
Supplier: LGC Standards PROMOCHEM
NIST-1546A
PROONIST-1546AEA 2310 EUR
PROONIST-1546A
Meat homogenate - nutrients, NIST-1546A
Standards Food Standards
This standard reference material is intended for determining fatty acids, cholestrol, proximates, calories, elements, vitamins and amino acids in canned meat products and similiar materials. The meat homogenate is a mixture of pork and chicken products blended together in a commercial process.
Certifications: Traceable to NIST and SRM numbers.