Copper standard solution, NIST-3114
Supplier: LGC Standards PROMOCHEM
NIST-3114
PROONIST-3114EA 1980 EUR
PROONIST-3114
Copper standard solution, NIST-3114
Standards Cationic Standards Copper Standards
This reference standard solution is intended for use as a primary calibration standard for the quantitative determination of copper.
A unit consists of 50 ml of an aqueous solution in a high-density polyethylene bottle sealed in an aluminised bag.
Certifications: Traceable to NIST and SRM numbers.