Organic reference standards, Vamidothion sulphone (NMIA P1730)

Supplier: LGC Standards PROMOCHEM

NMIAP1730
PROONMIAP1730EA 483 EUR
PROONMIAP1730
Organic reference standards, Vamidothion sulphone (NMIA P1730)
Standards Organic Standards

Purity estimate obtained by subtraction from 100% of total impurities by HPLC with UV detection, thermogravimetric analysis, Karl Fischer analysis and 1H NMR analysis is 96,1 ± 4,0% mass fraction, (95% coverage interval). Supporting evidence is provided by elemental microanalysis and quantitative NMR using certified triphenylphosphine oxide as internal standard.


  • White solid
  • Moisture content ≤0,2% mass fraction
  • Volatile residue ≤0,1% mass fraction
  • Non volatile residue ≤0,4% mass fraction
  • CAS no.: 70898-34-9


Synonym: O,O-dimethyl-S-[2-[(1-methyl-2-(methylamino)-2-oxoethyl) sulphonyl] ethyl] phosphorothioate.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR