Nitrite ion chromatography standard, VHG
Supplier: LGC Standards PROMOCHEM
VHG-INO2-100 VHG-INO2-500
PROOVHG-INO2-100EA 62.1 EUR
PROOVHG-INO2-100 PROOVHG-INO2-500
Nitrite ion chromatography standard, VHG
Standards Anionic Standards Nitrite Standards
This solution is intended for use as a certified reference material (CRM) calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and SRM.