Tin standard solution, for ICP, in HCl, CRM, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PSNH-100 VHG-LSNH-100 VHG-PSNH-50 VHG-PSNH-500 VHG-TSNH-100 VHG-TSNH-500
PROOVHG-PSNH-100EA 54.6 EUR
PROOVHG-PSNH-100 PROOVHG-LSNH-100 PROOVHG-PSNH-50 PROOVHG-PSNH-500 PROOVHG-TSNH-100 PROOVHG-TSNH-500
Tin standard solution, for ICP, in HCl, CRM, VHG
Standards Cationic Standards Tin Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.