Yttrium standard solution, for ICP, in HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PYN-100 VHG-TYN-100 VHG-PYN-250 VHG-LYN-100 VHG-PYN-50 VHG-PYN-500 VHG-TYN-500
PROOVHG-PYN-100EA 53.5 EUR
PROOVHG-PYN-100 PROOVHG-TYN-100 PROOVHG-PYN-250 PROOVHG-LYN-100 PROOVHG-PYN-50 PROOVHG-PYN-500 PROOVHG-TYN-500
Yttrium standard solution, for ICP, in HNO₃, CRM, VHG
Standards Cationic Standards Yttrium Standards
These standard solution are intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF) and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.