Gold standard solution, for ICP, in 20% HCl, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PAUH-100 VHG-PAUH-50 VHG-PAUH-500 VHG-TAUH-100 VHG-TAUH-500
PROOVHG-PAUH-100EA 107 EUR
PROOVHG-PAUH-100 PROOVHG-PAUH-50 PROOVHG-PAUH-500 PROOVHG-TAUH-100 PROOVHG-TAUH-500
Gold standard solution, for ICP, in 20% HCl, VHG
Standards Cationic Standards Gold Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.