Ytterbium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PYBN-100 VHG-PYBN-500 VHG-TYBN-100 VHG-TYBN-500
PROOVHG-PYBN-100EA 65.3 EUR
PROOVHG-PYBN-100 PROOVHG-PYBN-500 PROOVHG-TYBN-100 PROOVHG-TYBN-500
Ytterbium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards Cationic Standards Ytterbium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.